Probes, Very Fine Tip (0.001"), Stainless Steel, Excelta Corp®
Supplier: Excelta
Certificates
About this item
Smallest tip made - great for magnification and very fine work.
- Extreme fine tips for extreme fine work
- Available in short or long handle
- Corrosion resistant
These stainless steel probes have a 1 mil tip. The 335 Series Mini-Probes are 4.75" long. The 332 series are 6.5" long. They feature the smallest, finest tips available on any probe and are ideal for microscopy.
Frequently Bought Together