Synthware Low Form Dewar Flasks, Hemispherical, Kemtech America
Fournisseur: Kemtech America
Certificats
À propos de cet article
Flask size shown is the maximum capacity flask that will fit inside the dewar.
- Heavy wall borosilicate inner construction
- Aluminum housing
- Weighted metal base
Hemispherical dewars work great with flasks up to the size listed.
Spécifications
- Catalog No:
- 60001-188
- 60001-186
- 60001-184
- Capacity:
- 0.5 L
- 0.25 L
- 0.1 L
- Int. height:
- 65 mm
- 55 mm
- 40 mm
- Ext.Ø:
- 140 mm
- 100 mm
- 80 mm