Double-Blade Squeegees, New Pig
Supplier: New Pig
Certificates
About this item
Chemical-resistant PVC blades leave floors free of liquid and streaks.
- Solid, 60" handle keeps users a safe distance from spill
- Non-sparking, easy-to-clean PVC won't rust or corrode
Two blades double the service life because the head flips, and the unused blade acts as a splashguard.
Ordering information: Chemical-resistant PVC blades and 60" handle are sold separately.
Frequently Bought Together