ContactUsLinkComponent
Twist Mops, Micronova
Catalog # 68400-248
Supplier:  Avantor
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
   Waste Reducing
undefined
Twist Mops, Micronova
Catalog # 68400-248
Supplier:  Avantor
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
   Waste Reducing
Specifications
  • Description:
    Twist Mop Head, PVA
  • Environmentally Preferable:
    Waste Reducing
  • Cat. no.:
    68400-248
  • Supplier No.:
    TM-1
Specifications
About this item
Mop system cuts cleaning time and eliminates unnecessary hardware by incorporating a simple self-wringing action in the handle
The mop can wipe and clean a floor to a virtual dry finish or apply a moist layer of sterilant to self dry. Effectively absorbs liquid spills. Mop head is available in two different fabrics for varying cleanroom demands. The aluminum and nylon handle and polyester mop head are autoclavable.
Ordering information: Mop heads and handle are ordered separately.