BCR® Mops, Berkshire
47729-382
Small Business Enterprise
- Description:Mop 4
- Material:Knitted Polyester
- Size:2
- Cat. no.:47729-382
- Supplier No.:BCM.4.12
Durable, absorbent mops designed for the easy removal of contamination from floors and for the distribution of disinfectants.
- Soft, non-abrasive
- Disposable
- Economic and highly absorbent
- Autoclavable
- ISO Class 5–8
- Cleanroom Class 100–100,000 (Mop 1), 1–100,000 (Mop 4)
All components of the mopping system may be autoclaved or irradiated.
Mops feature a fan-tail design and looped-end construction to minimize particle generation. Mop 1 is made of nonwoven polyester, and is soft and nonabrasive. Mop 4 is made from continuous filament tubular knitted polyester and is chemical- and abrasion-resistant. Mop 4 has low particulate and fibers, a lot number to assure traceability, and is available sterile. Mop 4 is laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10) cleanroom.