MegaTip™ Swab Mops, Micronova
The MegaTip™ is a substantial cleaning swab that will easily reach to clean recesses, corners, and angles between walls, ceilings, and other hard to reach areas. When used with the SSU series handle, MegaTip™ Swab Mops may be used for applications where mop or wipe use is not practical.
- Polyester/foam swab tip for cleaning 4×1.5" (10×4cm) areas
- Stainless steel adapter
- Available in smooth (NovaPoly) or textured (PolySorb) polyester formats
- Compatible with alcohol and strong disinfectants
- Available gamma irradiated for aseptic areas and equipment
The foam tip swab slips onto the MegaTip™ stainless steel frame and adapter. The gamma irradiated polyester covered swabs are ideal for aseptic cleanrooms.
Frequently Bought Together