Wall WipR™ Cleaning System, Contec® | Avantor
Wall WipR™ Cleaning System, Contec®
Supplier:   Contec
Wall WipR™ Cleaning System, Contec®
Supplier:  Contec
Certificates
Wall WipR™ Cleaning System, Contec®
2 Options Available Below
Product Details & Documents
Designed for easy, effective cleaning of walls, ceilings and other flat surfaces in controlled environments
  • Economical, effective and efficient cleaning system for vertical surfaces
  • Convenient 14" (35.6 cm) and 18" (45.7 cm) sizes
  • Fabric covers are low in extractable residues, and even lower in particle generation
Swivel T-handles, aluminum extension poles, and wiping covers are available in two sizes. Casters for the plastic bucket are available for system mobility.
System consists of a swivel T-handle with a left-right pivot, an extension pole attached to the handle, fabric wiping covers with laminated foam cushions, and a bucket with wringer sieve. Wiping covers are lightly stretched over the T-handle, providing a wide wiping path and maneuverability for hard-to-reach areas. Handle and covers are low in extractable residues.
Ordering information: All components must be ordered separately.
Packaging: Wall WipR™ covers are packaged 50 per bag, 8 bags per case. Wall WipR™ bucket casters are supplied as a set of four.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Availability
    Price
  • Wall WipR™ Polyester/Foam Wiping Cover
    35.6 cm (14") Wide
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $251.12
    $251.12
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 400
    Case of 400 - $2,008.96
    $2,008.96
    Prix promotionnel pour les nouveaux clients
    Pack of 50
       $251.12
    Case of 400
       $2,008.96
    Supplier Number: WWC14
  • Wall WipR™ Polyester/Foam Wiping Cover
    45.7 cm (18") Wide
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $251.98
    $251.98
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 400
    Case of 400 - $2,015.84
    $2,015.84
    Prix promotionnel pour les nouveaux clients
    Pack of 50
       $251.98
    Case of 400
       $2,015.84
    Supplier Number: WWC18
  • Wall WipR™ Polypropylene Bucket, Blue
    Discontinued
    Supplier Number: -