About this item
The Model 9100 storage unit is engineered and designed for the secure storage and organization of microscope slides and paraffin embedded cassettes within a single unit.
- Secure storage
- Stackable
Each storage unit features robust uniblock construction with minimized spot welding for increased support.
An organizer tray can be positioned on top of the unit for use as a workspace or for prepping drawers. Index tabs and drawer labels are available for quick identification and sorting. The optional cabinet base is equipped with four casters for mobility. The front two casters are lockable.
Stackable layers of slide and cassette drawers allow for customizable storage, with a possible total of 14 slide drawers and 8 cassette drawers. All drawers feature a finger pull for easy removal and insertion. Drawer interiors can be accessed easily due to plastic coated rail inserts that facilitate opening and closing, whether or not the drawers are loaded.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.