Spectra/Por® Magnetic Weighted Closures, Spectrum® Laboratories
Supplier: Spectrum Laboratories
Certificates
About this item
Closures feature imbedded fluoropolymer resin-coated magnetic weights
Sink in water. Provide a slow spin movement of dialysis membrane when used with a magnetic stir plate. No additional stirring bar is required to maintain continuous motion. Not for use with cellulose ester (CE) or polyvinylidene fluoride (type F) tubing.
Frequently Bought Together