Tissue Marking Dyes (TMD™), TBS®
Tissue Marking Dyes (TMD™), TBS®
Tissue Marking Dyes (TMD™), TBS®
Certificates
Tissue Marking Dyes (TMD™), TBS®
Tissue Marking Dyes (TMD™), TBS®
6 Options Available Below
Product Details & Documents
Ideal for marking margins or for processing multiple specimens in one tissue cassette, these dyes have been reformulated for superior adherence to tissue surfaces
Viscosity allows a thin, even coating of pigment without masking underlying tissue. Quick drying decreases frozen sectioning time and provides excellent readability. Size: 237 mL (8 oz.).
Product Family Options
Product Information
  • Désignation
    Color
    Taille
    Availability
    Price
  • Tissue Marking Dyes (TMD™)
    Black
    8 oz
    Discontinued
    Supplier Number: TMD-BK
  • Tissue Marking Dyes (TMD™)
    Yellow
    8 oz
    Discontinued
    Supplier Number: TMD-Y
  • Tissue Marking Dyes (TMD™)
    Orange
    8 oz
    Log in for availability
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $608.17
    $608.17
    Log in for availability
    Case of 4 (1/ Itm/Bx)
    Case of 4 (1/ Itm/Bx) - $2,432.68
    $2,432.68
    Each(1/Itm/Bx)
       $608.17
    Case of 4 (1/ Itm/Bx)
       $2,432.68
    Supplier Number: TMD-O
  • Tissue Marking Dyes (TMD™)
    Black
    8 oz
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $556.89
    $556.89
    Log in for availability
    Case of 4
    Case of 4 - $2,227.56
    $2,227.56
    Each
       $556.89
    Case of 4
       $2,227.56
    Supplier Number: TMD-BK
  • Tissue Marking Dyes (TMD™)
    Blue
    8 oz
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $556.89
    $556.89
    Log in for availability
    Case of 4
    Case of 4 - $2,227.56
    $2,227.56
    Each
       $556.89
    Case of 4
       $2,227.56
    Supplier Number: TMD-BL
  • Tissue Marking Dyes (TMD™)
    Yellow
    8 oz
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $549.05
    $549.05
    Log in for availability
    Case of 4
    Case of 4 - $2,196.20
    $2,196.20
    Each
       $549.05
    Case of 4
       $2,196.20
    Supplier Number: TMD-Y
  • Tissue Marking Dyes (TMD™)
    Red
    8 oz
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $556.89
    $556.89
    Log in for availability
    Case of 4
    Case of 4 - $2,227.56
    $2,227.56
    Each
       $556.89
    Case of 4
       $2,227.56
    Supplier Number: TMD-R
  • Tissue Marking Dyes (TMD™)
    Green
    8 oz
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $556.89
    $556.89
    Log in for availability
    Case of 4
    Case of 4 - $2,227.56
    $2,227.56
    Each
       $556.89
    Case of 4
       $2,227.56
    Supplier Number: TMD-G