Germicidal Bulbs
Germicidal Bulbs
Supplier:   Bulbtronics
Germicidal Bulbs
Supplier:  Bulbtronics
Certificates
Germicidal Bulbs
Germicidal Bulbs
9 Options Available Below
Product Details & Documents
Various germicidal bulbs.
Product Family Options
Informations sur le produit
  • Désignation
    Modèle
    Puissance
    Wavelength
    Fond
    Bulb Life
    Longueur
    Ø
    Availability
    Price
  • Bulb, Linear Double Ended, General Electric, Preheat (Circuitry)
    T5
    6 W
    253.7 nm
    G5/mini 2-pin
    6000 h
    203.2 mm
    16 mm
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    80094-318
  • Bulb, Linear Double Ended, General Electric, Preheat (Circuitry)
    T8
    25 W
    253.7 nm
    G13/medium 2-pin
    7500 h
    457.2 mm
    25.4 mm
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    80094-308
  • Bulb, Linear Double Ended, General Electric, Preheat (Circuitry)
    T5
    39 W
    253.7 nm
    FA8/slimline, single pin
    9000 h
    914.4 mm
    16 mm
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    80094-312
  • Bulb, Linear Double Ended, Light Sources
    T8
    10 W
    253.7 nm
    G13/medium 2-pin
    7500 h
    342.9 mm
    25.4 mm
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-151
  • Bulb, Linear Double Ended, Light Sources, Ceramic Base Germicidal
    T5
    25 W
    253.7 nm
    FA8/slimline, single pin
    9000 h
    558.8 mm
    15.875 mm
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-281
  • Bulb, Linear Double Ended, Light Sources, Rapid Start (Circuitry)
    T5
    39 W
    253.7 nm
    FA8/slimline, single pin
    7500 h
    843.28 mm
    15.875 mm
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-183
  • Bulb, Light Sources, Linear Double Ended, Rapid Start (Circuitry)
    T5
    65 W
    253.7 nm
    FA8/slimline, single pin
    7500 h
    1574.8 mm
    15.875 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $309.04
    $309.04
    Each
       $309.04
    Supplier Number: 0000187
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-214
  • Bulb, Norman Lamps, Linear Double Ended, Preheat; Starter Required (Circuitry)
    T5
    8 W
    253.7 nm
    G5/mini 2-pin
    7500 h
    298.4 mm
    15.875 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $49.87
    $49.87
    Each
       $49.87
    Supplier Number: 0000194
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-228
  • Bulb, Linear Double Ended, Osram Sylvania, Preheat (Circuitry); Soft Glass; Mineral-Light Puritec
    T5
    4 W
    253.7 nm
    G5/mini 2-pin
    8000 h
    152.4 mm
    15.875 mm
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    89174-746
  • Bulb, Linear Double Ended, Philips, Disinfection Water Air Treatment
    T5
    40 W
    253.7 nm
    FA8/slimline, single pin
    9000 h
    863.6 mm
    15.875 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $239.50
    $239.50
    Each
       $239.50
    Supplier Number: 0000234
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-308
  • Bulb, Compact, Philips, Hot Cathode
    Twin Tube
    9 W
    253.7 nm
    G23/2-pin
    9000 h
    165.1 mm
    12.7 mm
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-300
  • Bulb, Linear Double Ended, Philips, Hot Cathode
    T8
    15 W
    253.7 nm
    G13/medium 2-pin
    8000 h
    457.2 mm
    25.4 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $245.45
    $245.45
    Each
       $245.45
    Supplier Number: 0000157
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-154
  • Bulb, Linear Double Ended, Philips, Hot Cathode
    T8
    30 W
    253.7 nm
    G13/medium 2-pin
    8000 h
    914.4 mm
    25.4 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $289.04
    $289.04
    Each
       $289.04
    Supplier Number: 0000233
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-306
  • Bulb, Linear Double Ended, Philips, Sterilamp Alto
    T5
    75 W
    253.7 nm
    FA8/slimline, single pin
    9000 h
    1574.8 mm
    15.875 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $300.42
    $300.42
    Each
       $300.42
    Supplier Number: 0014860
    Spécifications:
    More
    Specifications
    Cat. No.
    80078-846
  • Bulb, Linear Double Ended, Sankyo Denki
    T10
    20 W
    253.7 nm
    G13/medium 2-pin
    8000 h
    588.5 mm
    32.5 mm
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-166
  • Bulb, Linear Double Ended, Sankyo Denki, Rapid Start (Circuitry)
    T10
    40 W
    253.7 nm
    G13/medium 2-pin
    8000 h
    1198 mm
    32.5 mm
    Discontinued
    Supplier Number: 0000179
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-198
  • Bulb, Linear Double Ended, Ushio
    T8
    10 W
    253.7 nm
    G13/medium 2-pin
    8000 h
    330 mm
    25.4 mm
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-152
  • Bulb, Linear Double Ended, Ushio
    T5
    16 W
    253.7 nm
    FA8/slimline, single pin
    7500 h
    375 mm
    15 mm
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-140
  • Bulb, Linear Double Ended, Ushio, Preheat (Circuitry)
    T8
    30 W
    253.7 nm
    G13/medium 2-pin
    8000 h
    893 mm
    25.4 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $136.82
    $136.82
    Each
       $136.82
    Supplier Number: 0016451
    Spécifications:
    More
    Specifications
    Cat. No.
    80077-620
  • Bulb, Linear Double Ended, Ushio, Preheat; Universal Burn (Circuitry)
    T5
    7.2 W
    253.7 nm
    G5/mini 2-pin
    8000 h
    287 mm
    15.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $59.05
    $59.05
    Each
       $59.05
    Supplier Number: 0000195
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-230
  • Bulb, Linear Double Ended, Ushio, Universal Burn
    T5
    4.5 W
    253.7 nm
    G5/mini 2-pin
    6000 h
    134.5 mm
    15.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $54.50
    $54.50
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $545.00
    $545.00
    Each
       $54.50
    Pack of 10
       $545.00
    Supplier Number: 0000183
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-206
  • Bulb, Linear Single Ended, Voltarc
    T5
    16 W
    253.7 nm
    S400/4-pin
    7000 h
    357 mm
    15.875 mm
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-302
  • Bulb, Linear Double Ended, Voltarc, Instant Start (Circuitry)
    T5
    16 W
    253.7 nm
    FA8/slimline, single pin
    9000 h
    356 mm
    15.875 mm
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-145
  • Bulb, Linear Single Ended, Voltarc, Instant Start (Circuitry)
    T6
    39 W
    253.7 nm
    S406/4-pin with integral jumpers
    8000 h
    864 mm
    19.05 mm
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    47736-188