ContactUsLinkComponent
Chromacol Headspace Caps and Septa
Catalogue # 76181-356
Fournisseur:  Thermo Scientific
Chromacol Headspace Caps and Septa
Catalogue # 76181-356
Fournisseur:  Thermo Scientific
Supplier Number:  WSKTCL-100
Spécifications
  • Septum:
    PTFE/Silicone, slit
  • Description:
    Closures wash kit, PE snap cap T/S, slit, 100 per pack
  • Cat. no.:
    76181-356
Spécifications
À propos de cet article
Chromacol headspace caps and septa use magnetic seals with PAL/CTC/Leap Technologies, Gerstel and other magnetic transport autosamplers.
  • 20 mm crimp seals must be applied with a crimping tool
  • Pre-assembled caps and septa are convenient and minimize contamination from handling
  • Extended temperature range products for high-temperature headspace applications