Chromacol Headspace Caps and Septa
Catalogue # 76181-356
Fournisseur: Thermo Scientific
Spécifications
- Septum:PTFE/Silicone, slit
- Description:Closures wash kit, PE snap cap T/S, slit, 100 per pack
- Cat. no.:76181-356
Spécifications
À propos de cet article
Chromacol headspace caps and septa use magnetic seals with PAL/CTC/Leap Technologies, Gerstel and other magnetic transport autosamplers.
- 20 mm crimp seals must be applied with a crimping tool
- Pre-assembled caps and septa are convenient and minimize contamination from handling
- Extended temperature range products for high-temperature headspace applications