Analytical Mill, A 10 basic
A 10 basic
Catalog # 76329-858
Supplier: IKA Works
Supplier #: 0025002269
Jump to:
Product Details & Documents
This batch mill has simple handling with keypads which grinds hard, brittle, soft and fibrous materials, because samples may be embrittled directly in the grinding chamber- tough, oily and aqueous samples can also be grinded. Easily exchangeable beater/cutter for a variety of applications.
- Counter- displays grinding time and error code
- Digital timer with interval function, integrated cooling connections
- Brushless motor for long service life and low noise level
- Bayonet lock for lid to enable simple and safe locking
The mill starts only when the lid is closed and can be opened at standstill. A quick stop feature further increases the safety of the user.
Specifications
- Désignation:A 10 basic