Recirculating Chillers, RC 2 | Avantor
Recirculating Chillers, RC 2
Supplier:   IKA Works
Recirculating Chillers, RC 2
Supplier:  IKA Works
Certificates
Recirculating Chillers, RC 2
Recirculating Chillers, RC 2
2 Options Available Below
Product Details & Documents
Designed for cooling applications up to −20 °C with a powerful 400 W cooling capacity, these recirculating chillers are highly efficient and provide stable temperatures. With a wireless controller, it is possible to operate the RC 2 safely and comfortably irrespective of where the device is located.
  • A feed hopper bleed valve enables safe and clean handling of the thermofluid
  • Easy to open front cover allows the air filter to be conveniently cleaned
  • It is possible to measure and control temperature directly in the working medium via the external temperature sensor connection socket
Due to its outstanding temperature stability, the RC can handle even the most demanding applications. The speed-controlled PEEK pump allows an indirect continuous adjustment of the maximum pressure and flow rate.
The high quality insulated 4 L bath reservoir allows the use of up to 2.5 liters of thermofluid in the external cooling circuit without refilling.
Max. flow rate / max. pressure: 18 L/min, 0.3 bar.
 : Cooling capacity at +20; +10; 0; −10 and −20 °C.
Product Family Options
Informations sur le produit
  • Modèle
    Plage de température
    Capacité
    W×D×H
    Temperature Stability
    Availability
    Price
  • RC 2 basic
    −20...+40 °C
    0.4 kW
    220×450×478 mm
    ±0.05
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $12,446.45
    $12,446.45
    Each
       $12,446.45
    La quantité minimale de commande est de 1
    Supplier Number: 0004171001
  • RC 2 control
    −20...+40 °C
    0.4 kW
    220×450×478 mm
    ±0.05
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $17,611.53
    $17,611.53
    Each
       $17,611.53
    La quantité minimale de commande est de 1
    Supplier Number: 0004173001