Rotary Evaporators, RV 8
Fournisseur: IKA Works
Certificats
À propos de cet article
These entry-level evaporators are ideal for all standard evaporation applications in universities and research laboratories. The functions of the RV10 control have been extended and improved to enable fully automatic distillation: With automatic boiling point determination, an integrated solvent library and the option to connect a speed-controlled vacuum pump.
- All models have manual lift with safety stop function
- Digital TFT display showing the rotation speed and heating bath temperature allows optimal control of the entire distillation process
- Single-hand lift system with pressure sensors can be operated easily with the right or left hand
- Water/oil heating bath can be used as a stand-alone unit and has integrated ergonomic carrying handles
- New clamp mechanism for an easier changing of evaporation flasks
- USB connection enables online update of the firmware
Delivery: Units are supplied with heating bath HB 10, either with or without glassware. RV 8 models are compatible with the full RV10 glassware range.
Please note: A vacuum source, vacuum check valve and cooling system are required but not supplied. Please contact your local Avantor sales office for advice and details.
Please note: A vacuum source, vacuum check valve and cooling system are required but not supplied. Please contact your local Avantor sales office for advice and details.
Caution: * Flex package is delivered with bath, vapour tube, clamps for glassware and Woulf bottle only. This package enables the customer to select the glassware set up for their specific application.
Frequently Bought Together