Tissue Capsules, Simport Scientific
Tissue Capsules, Simport Scientific
Supplier:  Simport Scientific
Certificates
Tissue Capsules, Simport Scientific
Tissue Capsules, Simport Scientific
2 Options Available Below
Product Details & Documents
These polypropylene capsules are suitable for holding tissue samples during processing
The lids have a frosted marking area for sample identification and an open mesh area to facilitate fluid exchange. The whole surface of the base is also an open mesh. The lid snaps securely on the base, eliminating the risk of tissue loss during processing. Color: white.
Product Family Options
Informations sur le produit
  • Diameter ×H
    Availability
    Price
  • 38×10 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $562.67
    $562.67
    Case of 1,000
       $562.67
    Supplier Number: M471
  • 28×5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $557.22
    $557.22
    Case of 1,000
       $557.22
    Supplier Number: M470