Embedding Cassettes, Simport Scientific
Embedding Cassettes, Simport Scientific
Embedding Cassettes, Simport Scientific
Embedding Cassettes, Simport Scientific
These disposable plastic tissue cassettes are suitable for holding and identifying tissue samples in processing, embedding, and sectioning procedures
The cassettes fit securely in microtome chuck adapters. They are molded from a high-density acetal polymer that is totally resistant to the chemical action of histological solvents, except for clear cassette 82028-100, which is made of polypropylene. The cassettes are designed to accept standard metal lids and will keep specimens safe during processing. The 45° anterior, slanted writing surface accepts markings easily, permitting sample identification throughout all stages of embedding. The cassettes can also be used for automatic cassette labeling instruments.
 : Cassettes are packed in three dispenser boxes of 500 cassettes.
Product Family Options
  • Désignation
    Color
  • Embedding Cassettes
    White
    Case of 1,500
    Case of 1,500 - $360.44
    $360.44
    Case of 1,500
       $360.44
     : M480-2
  • Embedding Cassettes
    Pink
    Case of 1,500
    Case of 1,500 - $357.66
    $357.66
    Case of 1,500
       $357.66
     : M480-3
  • Embedding Cassettes
    Green
    Case of 1,500
    Case of 1,500 - $357.66
    $357.66
    Case of 1,500
       $357.66
     : M480-4
  • Embedding Cassettes
    Lilac
    Case of 1,500
    Case of 1,500 - $360.94
    $360.94
    Case of 1,500
       $360.94
     : M480-10
  • Embedding Cassettes
    Orange
    Case of 1,500
    Case of 1,500 - $360.94
    $360.94
    Case of 1,500
       $360.94
     : M480-11
  • Embedding Cassettes
    Aqua
    Case of 1,500
    Case of 1,500 - $333.41
    $333.41
    Case of 1,500
       $333.41
     : M480-12
  • Embedding Cassettes
    Yellow
    Case of 1,500
    Case of 1,500 - $333.41
    $333.41
    Case of 1,500
       $333.41
     : M480-5
  • Embedding Cassettes
    Blue
    Pack of 500
    Pack of 500 - $97.21
    $97.21
    Case of 1500
    Case of 1500 - $291.63
    $291.63
    Pack of 500
       $97.21
    Case of 1500
       $291.63
     : M480-6
  • Embedding Cassettes
    Peach
    Pack of 500
    Pack of 500 - $111.09
    $111.09
    Case of 1500
    Case of 1500 - $333.27
    $333.27
    Pack of 500
       $111.09
    Case of 1500
       $333.27
     : M480-7
  • Embedding Cassettes
    Gray
    Pack of 500
    Pack of 500 - $111.09
    $111.09
    Case of 1500
    Case of 1500 - $333.27
    $333.27
    Pack of 500
       $111.09
    Case of 1500
       $333.27
     : M480-9
  • Embedding Cassettes
    Clear
    Case of 1,500
    Case of 1,500 - $226.26
    $226.26
    Case of 1,500
       $226.26
     : M480-1
  • Embedding Cassettes
    Tan
    Case of 1,500
    Case of 1,500 - $226.26
    $226.26
    Case of 1,500
       $226.26
     : M480-8
Accessories
  •  
    Description
    Color
  • CA87002-382
    Embedding Cassettes
    Gray
    Case of 750 - $320.58
  •  
    Description
  • CA73960-014
    Metal Lid
    Pack of 25 - $181.14