ERGOMAT® Smooth Nitril ESD Nitrile Floor Mats, ERGOMAT
: Ergomat
This smooth version of the ERGOMAT® Nitril mat is ideal for harsh areas with chemicals, where trolleys or chairs are commonly used, and for areas where easy cleanability is a must.
- Smooth Nitril ESD mats are static dissipative for areas requiring static discharge
- Latex-free
These mats are typically used in electronics, microprocessor manufacturing, explosives, and chemical storage industries.
: Manufacturer's three-year warranty.
Frequently Bought Together