SWINGSETTE™ M515 Tissue Processing and Embedding Cassettes, Simport Scientific
Disposable plastic cassettes hold tissue specimens during processing, embedding, and storage
A special hinge holds the base and cover together and allows cassettes to be opened and closed as often as necessary. The cover can be removed and reinserted easily without the danger of specimen loss. Anterior surface is slanted at a 45° angle for use with leading labeling instruments. Three frosted writing surfaces. Cassette colors are non-cytotoxic and non-metallic.
They are molded from a special, high-density acetal polymer and are completely resistant to the chemical action of histological solvents. Cassettes keep specimens safely submerged in liquid while the efficient flow-through slots maximize fluid exchange and ensure proper drainage.
: Each case contains three dispenser boxes of 500 cassettes with the lids closed.
Disposable plastic cassettes hold tissue specimens during processing, embedding, and storage
A special hinge holds the base and cover together and allows cassettes to be opened and closed as often as necessary. The cover can be removed and reinserted easily without the danger of specimen loss. Anterior surface is slanted at a 45° angle for use with leading labeling instruments. Three frosted writing surfaces. Cassette colors are non-cytotoxic and non-metallic.
They are molded from a special, high-density acetal polymer and are completely resistant to the chemical action of histological solvents. Cassettes keep specimens safely submerged in liquid while the efficient flow-through slots maximize fluid exchange and ensure proper drainage.
: Each case contains three dispenser boxes of 500 cassettes with the lids closed.
Product Family Options
- Color
- : M515-2
- : M515-3
- : M515-4
- : M515-5
- : M515-6
- : M515-7
- : M515-8
- : M515-9
- : M515-10
- : M515-11
- : M515-12
- : M515-16
- : M515-17
- : M515-18