These disposable plastic cassettes hold tissue or biopsy specimens during processing and embedding as well as in storage
Made of a high-density acetal polymer, the cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents. The efficient 1mm square openings maximize fluid exchange and ensure proper drainage. Large labeling areas are located on three sides of the cassettes. The anterior writing area is slanted at a 45° angle to make the cassette more compatible with cassette labeling instruments. Colors are non-cytotoxic and nonmetallic.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.