Deep Ledge Utility Carts, Metro™ | Avantor
Deep Ledge Utility Carts, Metro™
Supplier:   Metropolitan Wire
Deep Ledge Utility Carts, Metro™
Supplier:  Metropolitan Wire
Certificates
Deep Ledge Utility Carts, Metro™
Deep Ledge Utility Carts, Metro™
7 Options Available Below
Product Details & Documents
Utility carts feature corrosion-proof polyurethane shelves with 7cm (23/4") deep ledges to contain products and spills
Smooth surfaces are designed to resist staining and wipe clean easily.
They have chrome-plated posts and an ergonomically-designed, chrome-plated handle. Carts are mounted on four resilient 10.2cm (4") diameter rubber casters. Three-shelf units have an easy-to-position center shelf that is adjustable in 2.5cm (1") increments. Each shelf can hold up to 68kg (150lbs.), and each unit can hold up to 181kg (400lbs.). Height: 104cm (41").
Certifications: NSF listed.
Ordering information: Carts are shipped knocked down and require no tools to assemble.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Load Capacity
    Longueur
    Largeur
    Shipping Weight
    Availability
    Price
  • Three-Shelf Unit
    Black
    400 lbs.
    32¾"
    21½"
    37.5 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $833.78
    $833.78
    Each
       $833.78
    Supplier Number: BC2030-3DBL
  • Two-Shelf Unit
    Black
    400 lbs.
    38¾"
    27"
    35 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $794.94
    $794.94
    Each
       $794.94
    Supplier Number: BC2636-2DBL
  • Three-Shelf Unit
    Black
    400 lbs.
    38¾"
    27"
    46.5 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $977.90
    $977.90
    Each
       $977.90
    Supplier Number: BC2636-3DBL
  • Three-Shelf Unit
    Grey
    400 lbs.
    32¾"
    21½"
    37.5 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $782.22
    $782.22
    Each
       $782.22
    Supplier Number: BC2030-3DG
  • Two-Shelf Unit
    Grey
    400 lbs.
    38¾"
    27"
    35 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $724.59
    $724.59
    Each
       $724.59
    Supplier Number: BC2636-2DG
  • Three-Shelf Unit
    Grey
    400 lbs.
    38¾"
    27"
    46.5 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $969.26
    $969.26
    Each
       $969.26
    Supplier Number: BC2636-3DG
  • Two-Shelf Unit
    Grey
    400 lbs.
    32¾"
    21½"
    29 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $630.35
    $630.35
    Each
       $630.35
    Supplier Number: BC2030-2DG