SWINGSETTE™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
SWINGSETTE™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
SWINGSETTE™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
Supplier:  Simport Scientific
Certificates
SWINGSETTE™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
SWINGSETTE™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
28 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These cassettes are made of a high-density acetal polymer and keep specimens safely submerged in liquid
The cassettes are totally resistant to the chemical action of histological solvents. The flow-through openings allow for maximum fluid exchange and ensure proper drainage. Lids can be opened and closed as often as necessary and always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassettes. The anterior writing area is at a 45° angle. All cassettes colors are non-cytotoxic and non-metallic. Dimensions: 42.2Lx28.5Wx6Hmm (121/32x11/8x1/4").
Ordering information: Each case contains 10 sleeves of 75 bases and 10 bags of 75 lids. Cassette bases are packaged in transparent sleeves for quick loading into automatic labeling instruments.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Availability
    Price
  • Tissue Cassettes
    White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $298.04
    $298.04
    Case of 750
       $298.04
    Supplier Number: M517-2SL
  • Tissue Cassettes
    Pink
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $300.06
    $300.06
    Case of 750
       $300.06
    Supplier Number: M517-3SL
  • Tissue Cassettes
    Green
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $300.84
    $300.84
    Case of 750
       $300.84
    Supplier Number: M517-4SL
  • Tissue Cassettes
    Yellow
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.86
    $302.86
    Case of 750
       $302.86
    Supplier Number: M517-5SL
  • Tissue Cassettes
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $300.84
    $300.84
    Case of 750
       $300.84
    Supplier Number: M517-6SL
  • Tissue Cassettes
    Peach
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.86
    $302.86
    Case of 750
       $302.86
    Supplier Number: M517-7SL
  • Tissue Cassettes
    Tan
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.86
    $302.86
    Case of 750
       $302.86
    Supplier Number: M517-8SL
  • Tissue Cassettes
    Gray
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.86
    $302.86
    Case of 750
       $302.86
    Supplier Number: M517-9SL
  • Tissue Cassettes
    Lilac
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $446.40
    $446.40
    Case of 750
       $446.40
    Supplier Number: M517-10SL
  • Tissue Cassettes
    Orange
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $446.40
    $446.40
    Case of 750
       $446.40
    Supplier Number: M517-11SL
  • Tissue Cassettes
    Aqua
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.86
    $302.86
    Case of 750
       $302.86
    Supplier Number: M517-12SL
  • Tissue Cassettes
    Fluorescent Pink
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.87
    $302.87
    Case of 750
       $302.87
    Supplier Number: M517-16SL
  • Tissue Cassettes
    Fluorescent Green
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.87
    $302.87
    Case of 750
       $302.87
    Supplier Number: M517-17SL
  • Tissue Cassettes
    Fluorescent Yellow
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.87
    $302.87
    Case of 750
       $302.87
    Supplier Number: M517-18SL
  • Biopsy Cassettes
    White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.86
    $302.86
    Case of 750
       $302.86
    Supplier Number: M518-2SL
  • Biopsy Cassettes
    Pink
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.86
    $302.86
    Case of 750
       $302.86
    Supplier Number: M518-3SL
  • Biopsy Cassettes
    Green
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.86
    $302.86
    Case of 750
       $302.86
    Supplier Number: M518-4SL
  • Biopsy Cassettes
    Yellow
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.86
    $302.86
    Case of 750
       $302.86
    Supplier Number: M518-5SL
  • Biopsy Cassettes
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.86
    $302.86
    Case of 750
       $302.86
    Supplier Number: M518-6SL
  • Biopsy Cassettes
    Peach
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.86
    $302.86
    Case of 750
       $302.86
    Supplier Number: M518-7SL
  • Biopsy Cassettes
    Tan
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.86
    $302.86
    Case of 750
       $302.86
    Supplier Number: M518-8SL
  • Biopsy Cassettes
    Gray
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.86
    $302.86
    Case of 750
       $302.86
    Supplier Number: M518-9SL
  • Biopsy Cassettes
    Lilac
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.86
    $302.86
    Case of 750
       $302.86
    Supplier Number: M518-10SL
  • Biopsy Cassettes
    Orange
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.86
    $302.86
    Case of 750
       $302.86
    Supplier Number: M518-11SL
  • Biopsy Cassettes
    Aqua
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.86
    $302.86
    Case of 750
       $302.86
    Supplier Number: M518-12SL
  • Biopsy Cassettes
    Fluorescent Pink
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.87
    $302.87
    Case of 750
       $302.87
    Supplier Number: M518-16SL
  • Biopsy Cassettes
    Fluorescent Green
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.87
    $302.87
    Case of 750
       $302.87
    Supplier Number: M518-17SL
  • Biopsy Cassettes
    Fluorescent Yellow
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 750
    Case of 750 - $302.87
    $302.87
    Case of 750
       $302.87
    Supplier Number: M518-18SL