UVP Converter Plates, Analytik Jena
Fournisseur: Analytik Jena CA
Certificats
À propos de cet article
UV to White Light Converter Plate measuring 21 x 26 cm, converts Analytik Jena transilluminator’s ultraviolet light to white light for viewing protein gels and Coomassie blue stained or silver stained media
Visi-Blue™ converter plate is designed to convert 302 nm UV light to 480 nm blue light for use with SYBR® Green, SYPRO® Orange and GFP stains. Surface is safety-fused into metal frame for durability.
Phosphored plexiglass uses a white diffuser to convert UV radiation. Plate housing is scratch-resistant metal. Handles on two sides allow easy transport.
Frequently Bought Together