Diamond-Plate Switchboard Mats, Wearwell®
Fournisseur: Wearwell
Certificats
À propos de cet article
High-performance, nonconductive vinyl matting designed to protect personnel in areas with dangerous high-voltage equipment
Meet ANSI/ASTM D-178-01 Type 2, Class II standard.
Unique diamond-embossed surface cleans easily and will not collect contaminants. Mats insulate against up to 30,000V of electricity. Recommended working voltage: 17,000V.
Frequently Bought Together