ContactUsLinkComponent
Infrared Heating and Drying Lamp, Table Model, Walter Stern
Infrared Heating and Drying Lamp, Table Model, Walter Stern
Catalog # 36547-009
Supplier:  Walter Stern
undefined
Infrared Heating and Drying Lamp, Table Model, Walter Stern
Catalog # 36547-009
Supplier:  Walter Stern
Supplier Number:  750-001
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Cat. no.:
    36547-009
  • Electrical:
    115 V
  • Supplier No.:
    750-001
Specifications
About this item
Provides a convenient and reliable source for near-infrared rays for heating and drying
A 250W tungsten filament drying lamp is used. The on/off switch is on the lamp base. Arm is offset to allow placement of evaporating dish or other object to be heated directly under lamp. Bulb housing is adjustable on the lamp arm.
Ordering information: Complete with one 250W clear bulb, cord, and plug.