Pellet Cups® Sample Cups, Chemplex®
Supplier: Chemplex
Jump to:
About this item
Compressible, tapered sample cups for converting powdered sample materials into self-supported briquettes.
- Available in plastic or aluminum
- Aluminum cups are taller than plastic cups and capable of accepting greater volumes of sample materials
The briquetted powdered samples formed are durable for safe handling, firm and resistant to fissure development, and reinforced on all sides except for the surface to be analyzed.
5 Options Available Below
Product Details & Documents
Compressible, tapered sample cups for converting powdered sample materials into self-supported briquettes.
- Available in plastic or aluminum
- Aluminum cups are taller than plastic cups and capable of accepting greater volumes of sample materials
The briquetted powdered samples formed are durable for safe handling, firm and resistant to fissure development, and reinforced on all sides except for the surface to be analyzed.
Product Family Options
Product Information
- DésignationØAvailabilityPrice
- Supplier Number: 552
- Supplier Number: 535
- Supplier Number: 553
- Supplier Number: 545
- Supplier Number: 554