Combined Form, Mercury-Free Hydrometers, API, G&W Instruments
These petroleum hydrometers have a thermometer in the body
Thermometers are filled with organic red liquid and contain no mercury.
They measures specific gravity according to API degree scale. Approximate length: 380mm. Thermometer range: 0 to 150°F. Temperature of standardization: 60°F.
Frequently Bought Together