SP Bel-Art Adjustable Splash Shields, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
Ergonomically shaped acrylic shields with handles are easily adjusted horizontally and vertically. Shields can be pulled close to the body for a comfortable working position. Available in four different styles to fit any bench space or work style.
- Ergonomically shaped, handles are easily adjusted horizontally and vertically
- Can be pulled close to the body for comfortable working position
- Available in four different styles to fit any bench space or work style
Wall style has a 61cm (24") long nonsagging arm; island bench style has 96.5cm (38") long nonsagging arm
Attachment accessories are ordered separately. Weighted base adapter 16667-104 is for use with shield 16667-108 only.
Double shields are supported by two arms. Single shield dimensions: 30.5Hx38.1Wcm (12x15"). Double shield dimensions: 38.1Hx61Wcm (15x24").
Frequently Bought Together