SP Bel-Art Large Weighted Safety Shields, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
Personnel are protected by a clear polycarbonate barrier with exceptional impact resistance and optical clarity.
- 4.7 mm (3/16") clear polycarbonate barrier
- Exceptional impact resistance and optical clarity
Outside base dimensions are 50W x 17D cm (191/2 x 61/2"). This item ships oversize.
Certifications: Meets impact-resistance requirements of ANSI Z97.1.
Spécifications
- Catalog No:
- 47750-422
- 13197-366
- Overall dimensions:
- 50 L x 17 W, cm (191/2 x 61/2")
- 50 L x 17 W, cm (191/2 x 61/2")