Exposure Control Plan Binder, Brady®
Exposure Control Plan Binder, 300, Black/orange on white, Polyethylene, 2", 11"
Catalog # CA56606-560
Supplier: Brady Worldwide
Fournisseur #: BH2023
Jump to:
Product Details & Documents
Exposure control plan binder to keep exposure control plan organized and protected.
- Ring size 1.5"
Binder comes with an attached 36" chain and a pair of sheet lifters.
Ordering information: Includes binder only, information center and accessories sold separately.
Spécifications
- Sheet Capacity:300
- Color:Black/orange on white
- Désignation:Exposure Control Plan Binder
- Hauteur:11"
- Matière:Polyethylene
- Profondeur:2"