Drum Thieves, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
About this item
Conventional open-end drum thieves
Insert empty tube in drum, hold your thumb over top and withdraw. Soda-lime glass units easily broken for quick disposal. All are 1.09 m long.
2 Options Available Below
Product Details & Documents
Conventional open-end drum thieves
Insert empty tube in drum, hold your thumb over top and withdraw. Soda-lime glass units easily broken for quick disposal. All are 1.09 m long.
Product Family Options
Product Information