Synthware Sublimators, Kemtech America
Supplier:   Kemtech America
Synthware Sublimators, Kemtech America
Supplier:  Kemtech America
Certificates
Synthware Sublimators, Kemtech America
Synthware Sublimators, Kemtech America
3 Options Available Below
Product Details & Documents
A grease-free, two-piece sublimation apparatus consisting of an upper water cooled condenser and a lower vessel to hold the material to be sublimed.
  • Heavy wall Borosilicate construction
  • Horseshoe clamp included
  • Grease free
  • Hi-Vac Valve included
The sublimate collects on the lower end of the condenser, which has a flat bottom. Condenser is constructed using a grease-free, threaded valve. The condenser and the lower vessel are connected by an O-ring joint, which enables the two parts to be gently separated for maximum product recovery. Complete apparatus consists of the cold finger condenser, lower vessel, Viton® O-ring, and stainless steel pinch clamp.
Product Family Options
Informations sur le produit
  • Capacité
    Joint Size
    Clamp Size
    O-Ring Size
    Availability
    Price
  • 10 ml (lower vessel)
    25
    35
    217
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $558.65
    $558.65
    Prix promotionnel pour les nouveaux clients
    Each
       $558.65
    Supplier Number: S732510
  • 40 ml (lower vessel)
    40
    65
    226
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $730.30
    $730.30
    Prix promotionnel pour les nouveaux clients
    Each
       $730.30
    Supplier Number: S734040
  • 60 ml (lower vessel)
    50
    75
    229
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $954.56
    $954.56
    Prix promotionnel pour les nouveaux clients
    Each
       $954.56
    Supplier Number: S735060
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Clamps for 35/20 spherical joints
    Clamps for 35/20 spherical joints
    60004-978
    Log in pour la disponibilité
    Each - $52.15
Informations sur le produit
  • Catalog #
    Availability
    Price
  • #65 stainless steel pinch clamp for #40 O-ring joint
    #65 stainless steel pinch clamp for #40 O-ring joint
    60004-982
    Log in pour la disponibilité
    Each - $137.34
Informations sur le produit
  • Catalog #
    I.D.×W
    O-Ring Size
    Availability
    Price
  • Viton® O-Ring
    Viton® O-Ring
    60005-024
    59.9 x 3.5 mm
    229
    Log in pour la disponibilité
    Each - $20.26
Informations sur le produit
  • Catalog #
    I.D.×W
    O-Ring Size
    Availability
    Price
  • Viton® O-Ring
    Viton® O-Ring
    60005-022
    50.4 x 3.5 mm
    226
    Log in pour la disponibilité
    Each - $17.48
Informations sur le produit
  • Catalog #
    Availability
    Price
  • #75 stainless steel pinch clamp for #50 O-ring joint
    #75 stainless steel pinch clamp for #50 O-ring joint
    60004-984
    Log in pour la disponibilité
    Each - $269.54