Synthware Column, Distilling, Vigreux, Microscale, Kemtech America
Catalogue # 60004-120
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- ST Joint No.:14/10
- Cat. no.:60004-120
- Indentation Length:75 mm
- Supplier No.:C221410B
Spécifications
À propos de cet article
Distilling column with multiple indents.
- Heavy wall borosilicate construction
- Ground glass joints
- Threaded upper joint
- Cap included
- Vigreux indentations
For microscale applications. Supplied with a nylon compression cap and Viton® O-ring.