Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Synthware Condenser, Reflux, Jacketed, Threaded, Microscale, Kemtech America

14/10, 80 mm, 8 mm
Catalog # 60004-124
Supplier:   Kemtech America
 :  C201080
Synthware Condenser, Reflux, Jacketed, Threaded, Microscale, Kemtech America
Catalog # 60004-124
Supplier:  Kemtech America
Synthware Condenser, Reflux, Jacketed, Threaded, Microscale, Kemtech America
Synthware Condenser, Reflux, Jacketed, Threaded, Microscale, Kemtech America

14/10, 80 mm, 8 mm

Price
$129.29
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
 
Product Details & Documents

Jacketed air reflux condenser with inner tube I.D

  • Heavy wall borosilicate construction
  • Ground glass joints
  • Jacketed

that is large enough to accept a thermometer. For microscale applications. Supplied with a nylon compression cap and Viton® O-ring.

Spécifications
  • ST Joint:
    14/10
  • Jacket Length:
    80 mm
  • Hose Connection O.D.:
    8 mm
Accessories

Informations sur le produit

  • Catalog #
    I.D.×W
    O-Ring Size
    Availability
    Price
  • Viton® O-Ring, 14/10 threaded joints
    Viton® O-Ring, 14/10 threaded joints
    60004-998
    014
    Log in pour la disponibilité
    Each - $5.61

Informations sur le produit

  • Catalog #
    Material
    Cap Size
    Availability
    Price
  • Nylon cap
    Nylon cap
    60005-054
    Nylon
    20 mm
    Log in pour la disponibilité
    Each - $4.65