Order Entry
Canada
ContactUsLinkComponent
Rubbermaid® Commercial 'Caution Wet Floor' Floor Sign
  89073-790
undefined
Rubbermaid® Commercial 'Caution Wet Floor' Floor Sign
  89073-790
 :  RCP611277YW

 

  • Color:
    Yellow
  • Height:
    25"
  • Material:
    Plastic
  • Cat. no.:
    89073-790
  • Depth:
    12"
  • Supplier No.:
    6112-77-YEL

 

 

The Rubbermaid Commercial 2-Sided Floor Safety Sign with "Caution Wet Floor" Imprint is lightweight and versatile.

  • Bright yellow plastic wet floor sign can be read from far away
  • 2-sided sign is ideal for doorways and narrow spaces
  • ANSI- and OSHA compliant color and graphics
  • Folds flat for storage or transport carts
  • Built-in carrying handle makes it easy to move and also clean
  • Measures 12" deep when open

The caution sign makes a heavy statement about safety. The 25-inch model is suitable for use in narrow areas. The folding design allows for convenient storage or transport on a janitor cart.

Each of the wet floor signs has caution warning messages imprinted in easy-to-read lettering for effective communication. The warning sign features durable plastic construction that will not rust, corrode or fade.

The flexible locking-style hanger makes the sign stationary and visible from all directions.