Rubbermaid® Commercial Commercial Multilingual 'Caution' Floor Sign
Catalog # 89073-248
Supplier: Janitorial Supplies
Specifications
- Color:Yellow
- Description:Folding Multilingual "Caution" Floor Sign
- Height:25"
- Material:Plastic
- Cat. no.:89073-248
- Legend:Caution;Attention;Cuidado
- Depth:12"
- Supplier No.:RCP 6112 YEL
Specifications
About this item
The Rubbermaid Commercial 2-Sided Caution Safety Sign is lightweight and versatile. The caution sign features the universal wet floor symbol and the word 'caution' imprinted in English, French and Spanish for effective communication to broad audiences.
- Folds easily for convenient storage or transport
- Two sided sign is ideal for doorways and narrow spaces
- Folds flat for storage or transport carts
- Built in carrying handle makes it easy to move and also clean
- Measures 12" deep when open
Each of the safety signs features durable plastic construction that will not rust, corrode or fade.
The flexible locking-style hanger makes the sign stationary and easily visible from all directions.
The 2-sided, bright yellow design meets OSHA requirements. ANSI compliant multi-lingual graphics warn of a nearby hazardous area