Saline Solutions, NERL Diagnostics
Saline Solutions, NERL Diagnostics
Saline Solutions, NERL Diagnostics
Certificates
Saline Solutions, NERL Diagnostics
Saline Solutions, NERL Diagnostics
2 Options Available Below
Product Details & Documents
Blood bank saline pH 7.0–7.2 is a phosphate buffered saline solution manufactured to match the pH of human blood
Manufactured to meet NCCLS standard ASI/BB-1. Certificates of analysis are available.
Note: Not for infusion, irrigation, or injection.
This preservative-free sodium chloride solution is for use in serological (blood bank) testing or for general laboratory use.
Documents
Product Family Options
Product Information
  • Volume
    Availability
    Price
  • 10 L (2.64 gal.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $593.30
    $593.30
    Log in for availability
    Pallet of 80 (1/ Itm/Bx)
    Pallet of 80 (1/ Itm/Bx) - $47,464.00
    $47,464.00
    Each(1/Itm/Bx)
       $593.30
    Pallet of 80 (1/ Itm/Bx)
       $47,464.00
    Supplier Number: 850510L
  • 20 L (5.3 gal.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $193.75
    $193.75
    Log in for availability
    Pallet of 48 (1/ Itm/Bx)
    Pallet of 48 (1/ Itm/Bx) - $9,300.00
    $9,300.00
    Each(1/Itm/Bx)
       $193.75
    Pallet of 48 (1/ Itm/Bx)
       $9,300.00
    Supplier Number: 8504