Washed Pooled Cells, Rockland Immunochemicals
Supplier: Rockland Immunochemical
Certificates
About this item
Suspensions are suitable for complement fixation, hemagglutination, hemadsorption, and other related serological procedures
All suspensions are 10% cell solutions in saline, except for RLR406-0050 and RLR406-0100. These two suspensions are 100% cells. Suspensions contain no stabilizers or preservatives.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together