Biofit Fixed-Height Lab Stools
Biofit Fixed-Height Lab Stools
Supplier:   BioFit
Biofit Fixed-Height Lab Stools
Supplier:  BioFit
Certificates
3 Options Available Below
Product Details & Documents
Durable fixed-height lab stools feature a steel seat attached to a steel plate, which is welded to a heavy-duty tubular base
Stools also have a fixed tubular footring, welded on both top and bottom for durability, and glides. Available in three colors.
Specifications for Product Family
Specifications
  • Catalog No:
  • BE-AJ2402
  • BE-AJ2404
  • BE-AJ2406
  • Model:
  • BE-AJ2402
  • BE-AJ2404
  • BE-AJ2406
  • Seat dimensions:
  • Ø13"
  • Ø13"
  • Ø13"
  • Shipping weight:
  • 11 lbs.
  • 11 lbs.
  • 11 lbs.
Specifications
Product Family Options
Product Information
  • Color
    Hauteur
    Foot Style
    Availability
    Price
  • Putty
    24"
    Glides
    Log in for availability
    Log in for availability
    Each
    Each - $1,096.82
    $1,096.82
    Each
       $1,096.82
    Supplier Number: AJ24-02
    Specifications:
    Product available on GSA Advantage®
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    BE-AJ2402
    Model
    BE-AJ2402
    Seat dimensions
    Ø13"
    Shipping weight
    11 lbs.
    Product available on GSA Advantage®
    Small Business Enterprise
  • Grey
    24"
    Glides
    Log in for availability
    Log in for availability
    Each
    Each - $1,111.67
    $1,111.67
    Each
       $1,111.67
    Supplier Number: AJ24-04
    Specifications:
    Product available on GSA Advantage®
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    BE-AJ2404
    Model
    BE-AJ2404
    Seat dimensions
    Ø13"
    Shipping weight
    11 lbs.
    Product available on GSA Advantage®
    Small Business Enterprise
  • Black
    24"
    Glides
    Log in for availability
    Log in for availability
    Each
    Each - $1,088.67
    $1,088.67
    Each
       $1,088.67
    Supplier Number: AJ24-06
    Specifications:
    Product available on GSA Advantage®
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    BE-AJ2406
    Model
    BE-AJ2406
    Seat dimensions
    Ø13"
    Shipping weight
    11 lbs.
    Product available on GSA Advantage®
    Small Business Enterprise