Microscope Slide Labels, TimeMed
Microscope Slide Labels, TimeMed
Certificates
Microscope Slide Labels, TimeMed
Microscope Slide Labels, TimeMed
3 Options Available Below
Product Details & Documents
This collection of labels provides an assortment of instrument-specific applications for microscope slides.
The film MicroTrax labels (36418-606) are for use with MicroTrax thermal transfer printers and run four across on a roll. The standard thermal transfer labels (36418-626 and -628) are for use with most thermal transfer printers, and are supplied in rows of four on a roll. All labels are white.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Dimensions
    Packaging
    Availability
    Price
  • Permanent microtrax labels
    White
    22.2×22.2 mm (0.88×0.88")
    7400 labels/roll, 3 rolls (22200 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 3
    Case of 3 - $878.47
    $878.47
    Case of 3
       $878.47
    Supplier Number: MICROTRXLBL4
  • Thermal transfer labels, glove friendly
    White
    22.2×22.2 mm (0.88×0.88")
    7400 labels/roll, 3 rolls (22200 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 3
    Case of 3 - $327.01
    $327.01
    Case of 3
       $327.01
    Supplier Number: TTR-7878
  • Thermal transfer labels, glove friendly
    White
    24×23.9 mm (0.94×0.94")
    7400 labels/roll, 3 rolls (22200 labels)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 3
    Case of 3 - $394.52
    $394.52
    Case of 3
       $394.52
    Supplier Number: TTR-1516