VWR® Biohazard Labels | Avantor
VWR® Biohazard Labels
VWR® Biohazard Labels
Supplier:  VWR International
Certificates
VWR® Biohazard Labels
VWR® Biohazard Labels
2 Options Available Below
Product Details & Documents
Labels aid in general workplace safety and efficiency by identifying contaminated equipment containers, rooms, or materials.
  • Quick and easy identification
  • Bold symbols declare biohazard warning from a distance
A biohazard symbol along with the term are printed in bold black atop a fluorescent red background.
Certifications: Labels are manufactured in accordance with various federal and state regulations, including OSHA safety standards.
Ordering information: Labels are available on rolls of 250 or 500 labels. One roll per case.
Documents
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Availability
    Price
  • Biohazard labels
    76.2×50.8 mm (3×2")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $47.54
    $47.54
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $47.54
  • Biohazard labels
    127×127 mm (5×5")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 250
    Case of 250 - $80.02
    $80.02
    Prix promotionnel pour les nouveaux clients
    Case of 250
       $80.02