À propos de cet article
Thermo Scientific Guardian Peroxidase Conjugate Stabilizer/Diluent preserves the functional integrity and activity of horseradish peroxidase conjugated antibodies and other proteins at very dilute concentrations for long-term storage.
- Preserves HRP activity: No significant loss of HRP activity over a six-month period at room temperature (1:1000 dilution) or 12 months at 4 °C
- Convenient: Store ready-to-use dilutions (1:1000 to 1:100,000) that maintain enzyme activity in the refrigerator—no aliquoting or freezing
- Assay compatible: Simply add blocking buffer to create the ideal diluent for your HRP-based ELISA system or store the HRP conjugate as a 1:1000 stock solution for Western blots and dilute in the final assay buffer
- Saves money: Less expensive than competitor formulations or ordering new HRP conjugates
With Guardian Peroxidase Conjugate Stabilizer/Diluent, typical 1 mg/mL antibody or streptavidin peroxidase conjugates can be diluted as much as 100,000-fold for storage at 4 °C. Most ELISA and blotting applications for HRP conjugates require dilution of typical 1 mg/mL stocks by at least 1000-fold (to 1 µg/mL). Sensitive assay systems, such as with chemiluminescent substrates, often require dilution up to 100,000-fold (10 ng/mL). For blotting applications, it is usually best to prepare a concentration for storage that can be diluted 10-fold in assay buffer (which may contain blocking components) for final use.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.