Glyco-SNAP-2
5 mg
CA80051-926
: 362339-5MG
A stable, water-soluble nitric oxide (NO) donor (1/2=27 hours) with properties similar to those of Glyco-SNAP-1 (80051-924). Dissolve in aqueous buffer immediately after use.
- Taille:5 mg
- Cat. No.:CA80051-926
- Enzyme name:Glyco-SNAP-2