Crews® Protective Goggles, MCR Safety
Fournisseur: MCR Safety
Certificats
À propos de cet article
Goggles provide a wide, unobstructed field of vision and are available in a variety of styles to protect against impact and splash.
- Wide, unobstructed field of vision
- Can be worn with most half-mask respirators and will fit comfortably over most prescription eyewear
- Transparent PVC frame and clear 0.06mm thick polycarbonate lens with scratch-resistant Duramass® coating
- Latex-free rubber straps provide secure fit on all styles
- Lenses provide 99.9% UV protection
- Goggles meet or exceed ANSI Z87.1+ safety standards
All goggles feature a transparent PVC frame and clear 0.06mm thick polycarbonate lens with scratch-resistant Duramass® coating. Latex-free rubber straps provide a secure fit on all styles. Lenses provide 99.9% UV protection.
They can be worn with most half-mask respirators and will fit comfortably over most prescription eyewear.
Certifications: Goggles meet or exceed ANSI Z87.1+ safety standards.
Frequently Bought Together