Standard Goggles, Sellstrom®
Goggles feature a flexible PVC body with 45.7 cm (18") long adjustable elastic headband.
Two venting options are available: indirect vent protects against liquids and dust, and direct vent provides maximum air flow with protection against grinding, chipping, and cutting operations. 1.5 mm (0.06") acetate lens available in clear or STA-CLEAR® anti-fog types.
These standard goggles have a green tinted body and fit comfortably over prescription glasses.
: Meet ANSI Z87.1-1989 standard.
Frequently Bought Together