TexNotes® Self-Adhesive Note Pads, Texwipe®
7.6 x 10.2 cm (3 x 4")
Catalog # TWTX5820
Supplier: Texwipe
Supplier #: TX5820
Jump to:
Product Details & Documents
Ideal for leaving notes on documents, reports, instruments, and most cleanroom surfaces
The low contamination, polymeric elastomer adhesive allows TexNotes to be attached and removed without high residue. Cleanroom packaged. 72 sheets/pad. Color: pastel blue
Specifications
- W×L:7.6 x 10.2 cm (3 x 4")