VWR® Red Border Tape Labels | Avantor
VWR® Red Border Tape Labels
VWR® Red Border Tape Labels
VWR® Red Border Tape Labels
VWR® Red Border Tape Labels
Designed to aid in the identification of containers, shelves, and other standard laboratory or workplace items.
  • Self-adhesive tape design
  • Distinctive coloring for quick identification
Labels are printed on a white. These labels can be written on with pen, pencil, or permanent marker. They can be easily removed and will not leave a sticky residue.
Ordering information: Supplied on individual rolls, each containing 12.7 m (41.7') of tape.
Product Family Options
  • Désignation
    Dimensions
  • Border tape labels
    25.4×19.1 mm (1×0.75")
    Case of 1
    Case of 1 - $22.77
    $22.77
    Case of 1
       $22.77
  • Border tape labels
    41.3×25.4 mm (1.63×1")
    Case of 1
    Case of 1 - $24.44
    $24.44
    Case of 1
       $24.44
  • Border tape labels
    63.5×38.1 mm (2.5×1.5")
    Case of 1
    Case of 1 - $25.30
    $25.30
    Case of 1
       $25.30
  • Border tape labels
    76.2×50.8 mm (3×2")
    Case of 1
    Case of 1 - $34.39
    $34.39
    Case of 1
       $34.39