Mounting Medium, Richard-Allan Scientific
Mounting Medium, Richard-Allan Scientific
Supplier:   Thermo Scientific
Mounting Medium, Richard-Allan Scientific
Supplier:  Thermo Scientific
Certificates
Mounting Medium, Richard-Allan Scientific
Mounting Medium, Richard-Allan Scientific
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Store slides for prolonged periods of time without fading of stains with Epredia™ mounting medium.
  • Rapid drying
  • Toluene-based
  • Optically clear
This toluene-based rapid drying mounting medium is miscible with all clearing agents including xylene substitutes. It is optically clear and will not cause fading of stains, even after prolonged storage. This mounting medium is miscible with all clearing agents, including xylene substitutes.
Ordering information: The 118.3 ml (4 oz.) bottle (84000-112) is a squeeze bottle with a twist-open/close cap.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • Mounting Medium
    473.2 mL (16 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2 (0.5 .pt US)
    Case of 2 (0.5 .pt US) - $536.45
    $536.45
    Case of 2 (0.5 .pt US)
       $536.45
    Supplier Number: 4111
  • Mounting Medium
    118.3 mL (4 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 6 (1/Itm/Bx)
    Case of 6 (1/Itm/Bx) - $273.64
    $273.64
    Case of 6 (1/Itm/Bx)
       $273.64
    Supplier Number: 4112