SC-3 System Hypalon® Sole Boots, WORKLON®
Supplier: Superior Uniform
À propos de cet article
Reusable, high-top cleanroom boots have a flexible Hypalon® sole with upper of static dissipative Burlington C3™ material
- Partial elastic opening with snap tab adjustment for a secure fit
- Rear snap attaches to the coverall
- Color-coded, buckled instep strap denotes size
Ordering information: Sole boots are sold in packs of one pair (2 pieces per pack).
5 Options Available Below
Product Details & Documents
Reusable, high-top cleanroom boots have a flexible Hypalon® sole with upper of static dissipative Burlington C3™ material
- Partial elastic opening with snap tab adjustment for a secure fit
- Rear snap attaches to the coverall
- Color-coded, buckled instep strap denotes size
Ordering information: Sole boots are sold in packs of one pair (2 pieces per pack).
Specifications for Product Family
Spécifications
- Catalog No:
- 14227-884
- 14227-886
- 14227-888
- 14227-890
- 14227-892
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Fabric:
- Hypalon®
- Hypalon®
- Hypalon®
- Hypalon®
- Hypalon®
Spécifications
Product Family Options
Informations sur le produit
- ColorPointureAvailabilityPrice
- Supplier Number: 1117-SSpécifications:
More
Specifications
Cat. No.14227-884Garment applicationCleanroomGenderUnisexFabricHypalon® - Supplier Number: 1117-MSpécifications:
More
Specifications
Cat. No.14227-886Garment applicationCleanroomGenderUnisexFabricHypalon® - Supplier Number: 1117-LSpécifications:
More
Specifications
Cat. No.14227-888Garment applicationCleanroomGenderUnisexFabricHypalon® - Supplier Number: 1117-XLSpécifications:
More
Specifications
Cat. No.14227-890Garment applicationCleanroomGenderUnisexFabricHypalon® - Supplier Number: 1117-2XLSpécifications:
More
Specifications
Cat. No.14227-892Garment applicationCleanroomGenderUnisexFabricHypalon®