FeTPPS
FeTPPS
25 mg
Catalog # CA80056-448
Supplier:   MilliporeSigma
Supplier #:  341492-25MG
FeTPPS
Catalog # CA80056-448
Supplier:  MilliporeSigma
CAS Number:  
FeTPPS
25 mg
CAS Number:  
Price
$335.90
Log in for availability
Each (25mg)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
A ferric porphyrin complex that catalytically isomerizes peroxynitrite to nitrate both in vivo and in vitro.
Provides cytoprotection against peroxynitrite (EC50=5 µM). Does not complex with nitric oxide and exhibits minimal SOD mimetic activity.
Specifications
Specifications
  • Taille:
    25 mg
  • Cat. No.:
    CA80056-448
  • Enzyme name:
    FeTPPS